Skip to Content
Merck
All Photos(1)

Key Documents

O3629

Sigma-Aldrich

Bis(2,4,6-trichlorophenyl) oxalate

Synonym(s):

2,4,6-Trichlorophenyl oxalate, Oxalic acid bis(2,4,6-trichlorophenyl) ester, TCPO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H4Cl6O4
CAS Number:
Molecular Weight:
448.90
Beilstein:
2312135
MDL number:
UNSPSC Code:
12352200

storage temp.

−20°C

SMILES string

Clc1cc(Cl)c(OC(=O)C(=O)Oc2c(Cl)cc(Cl)cc2Cl)c(Cl)c1

InChI

1S/C14H4Cl6O4/c15-5-1-7(17)11(8(18)2-5)23-13(21)14(22)24-12-9(19)3-6(16)4-10(12)20/h1-4H

InChI key

GEVPIWPYWJZSPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Useful in chemiluminescence enzyme-linked immunoassays.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemiluminescence enzyme immunoassay of 17 alpha-hydroxyprogesterone using glucose oxidase and bis(2,4,6-trichlorophenyl) oxalate - fluorescent dye system.
H Arakawa et al.
Chemical & pharmaceutical bulletin, 30(8), 3036-3039 (1982-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service