MTCR1020
MTC™ Bio Storage Box With Hinged Lid
to hold, 100 x 1.5 mL tubes, assorted colors
Synonym(s):
tube rack, tube storage box
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
assorted colors
polypropylene
feature
to hold 100 x 1.5 mL tubes
packaging
pack of 5 ea
manufacturer/tradename
MTC™ Bio R1020
Related Categories
General description
Storage Box, hinged lid, 100 x 1.5ml, Rainbow Pack of assorted colors, 5/pk
Features and Benefits
- Polypropylene, withstand -80°C
- 6 types - for 0.2mL to 50mL
- Compact and space efficient
- Autoclavable
Legal Information
MTC is a trademark of MTC Bio
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service