Key Documents
MTCP6420
MTC Bio ProPette Manual Pipette Controller
with filter
Synonym(s):
pipettor
Select a Size
1 EA
€83.20
€83.20
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€83.20
About This Item
UNSPSC Code:
41121500
NACRES:
NB.04
€83.20
Please contact Customer Service for Availability
packaging
pack of 1 ea
manufacturer/tradename
MTC Bio P6420
General description
ProPette MPC is the precise and easy-to-use manual pipette controller that‘s always ready to use. After attaching a pipette, one squeeze of its large rubber filling bulb is all it takes to start pipetting. There are no wires, no tubing, and no battery to recharge. Begin aspirating by touching the EZ-Flow lever, which is designed to precisely control the filling speed based on how gently or firmly you press the lever. A built-in replaceable filter ensures an optimum rate and protects the controller from liquid penetration and overfill. A separate button provides a positive blow-out function when required.
Features and Benefits
  • For glass or plastic pipettes
  • Accommodates pipettes up to 100mL
  • Easy, single lever control
  • Autoclavable lower body
  • Fingertip speed control
Legal Information
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)