MAK246
Neutrophil Elastase Activity Assay Kit (Fluorometric)
sufficient for 100 fluorometric tests
Synonym(s):
Elastase Enzyme Activity Fluorescent Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
detection method
fluorometric
relevant disease(s)
hematological disorder; pulmonary disorders
storage temp.
−20°C
General description
Neutrophil Elastase (NE, EC 3.4.21.37, leukocyte elastase, ELANE, ELA2, elastase 2, neutrophil elaszym, or serine elastase) is a cytotoxic serine protease. It is stored in the azurophil granules of neutrophil granulocyte and is released following cell stimulation, e.g. by pathogens, immune complexes or chemotactic agents (PMA). Mutation in the gene ELA2 (neutrophil elastase) results in neutropenia, a hereditary disorder characterized by reduced neutrophil count in an individual.
Application
- Relationship between airway inflammation and airflow limitation in elderly asthmatics.: This study investigates the connection between airway inflammation and airflow limitation in elderly asthmatic patients. It highlights the role of neutrophil elastase in airway inflammation, which was measured using the Neutrophil Elastase Activity Assay Kit (Fluorometric). The findings suggest that targeting neutrophil elastase activity could help in managing airflow limitation in elderly asthmatics, offering potential therapeutic applications for the assay kit (Soma et al., 2020).
Suitability
Suitable to determine the activity of purified Neutrophil Elastase, detect the activity of Neutrophil Elastase in plasma and/or blood and screen/study/characterize chemotactic agents causing Neutrophil stimulation and release of Elastase.
Principle
The Neutrophil Elastase Activity Assay Kit utilizes the ability of NE to proteolytically cleave a synthetic substrate and release a fluorophore, AFC, which can be easily quantified by fluorescence. This assay kit is simple, rapid, and can detect as low as 1 ng of neutrophil elastase in a variety of samples.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Neutrophil elastase in cyclic and severe congenital neutropenia.
Blood, 109(5), 1817-1824 (2007)
Bioresource technology, 302, 122791-122791 (2020-01-26)
The effect of co-cultivation of Porphyridium cruentum UTEX 161 with Pseudoalteromonas sp. MEBiC 03485 on P. cruentum growth and its sulfated polysaccharide (EPS) production were examined. The strain MEBiC 03485 had beneficial effects on P. cruentum growth, EPS production, and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service