Skip to Content
Merck
All Photos(1)

Key Documents

M2157

Sigma-Aldrich

Methyl β-L-daunosaminide hydrochloride

Synonym(s):

Daunosamine β-methyl glycoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO3 · HCl
CAS Number:
Molecular Weight:
197.66
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

form

powder

impurities

≤10% inorganic salts

SMILES string

Cl.COC1CC(N)C(O)C(C)O1

InChI

1S/C7H15NO3.ClH/c1-4-7(9)5(8)3-6(10-2)11-4;/h4-7,9H,3,8H2,1-2H3;1H

InChI key

JVYGBUAXDCLPMP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl β-L-daunosaminide hydrochloride has been used in a study to investigate enantiospecific synthesis of D-daunosamine from thioglycolic acid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantiospecific synthesis of D-daunosamine from thioglycolic acid
Po, S., et al.
Tetrahedron Asymmetry, 5, 1896-1872 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service