Skip to Content
Merck
All Photos(3)

Key Documents

KEM0032

Sigma-Aldrich

Terminal deoxynucleotidyl Transferase (TdT)

Ultra-pure enzyme for nucleic acid modifications

Sign Into View Organizational & Contract Pricing


About This Item

grade

for molecular biology

Assay

>99% (SDS-PAGE)

form

buffered aqueous solution

specific activity

27,400 U/mg

concentration

20,000 U/mL

shipped in

dry ice

storage temp.

−20°C

General description

Terminal deoxynucleotidyl transferase (TdT) is a template-independent DNA polymerase that catalyzes the addition of deoxynucleotides to the 3′ hydroxyl terminus of single or double stranded DNA molecules. The presence of 1 mM Co2+ stimulates the tailing of the 3′-ends of DNA fragments. This construct is sold as an N-terminal truncation of the terminal transferase gene attached to an N-terminal fusion tag.

Application

Suitable for:
  • Homopolymeric tailing to the 3′ OH
  • TUNEL assay
  • 5′-RACE
  • Labeling of 3′ ends

Features and Benefits

  • Ultra-purification process for ultimate enzyme performance
  • Highest quality specifications for ultimate product consistency
  • Undetectable DNA and nuclease contamination

Components

Supplied with:KEM0043B (10X Green Buffer)KEM0045B (2.5 mM CoCl2

Unit Definition

1 unit is defined as the amount of polymerase required to convert 1 nmol of dTTPs into acid insoluble material in 1 hour at 37° C.

Physical form

Supplied in 50 mM KPO4, 100 mM NaCl, 1.0 mM DTT, 0.1 mM EDTA, 0.1% Triton X-100, and 50% glycerol at pH 7.3 @ 25° C.

Other Notes

Source of protein: An E. coli strain that carries the cloned terminal transferase gene from calf thymus.
Unit size: 6,000 U

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laure Rittié et al.
Journal of cell communication and signaling, 2(1-2), 25-45 (2008-09-04)
Since molecular cloning has become routine laboratory technique, manufacturers offer countless sources of enzymes to generate and manipulate nucleic acids. Thus, selecting the appropriate enzyme for a specific task may seem difficult to the novice. This review aims at providing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service