Skip to Content
Merck
All Photos(3)

Key Documents

HPA007547

Sigma-Aldrich

Anti-ULBP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ALCAN-β antibody produced in rabbit, Anti-N2DL-1 antibody produced in rabbit, Anti-NKG2D ligand 1 precursor antibody produced in rabbit, Anti-NKG2DL1 antibody produced in rabbit, Anti-Retinoic acid early transcript 1I antibody produced in rabbit, Anti-UL16-binding protein 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

THCLCYDFIITPKSRPEPQWCEVQGLVDERPFLHYDCVNHKAKAFASLGKKVNVTKTWEEQTETLRDVVDFLKGQLLDIQVENLIPIEPLTLQARMS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ULBP1(80329)

General description

ULBP1 (Unique Long 16 binding protein 1) gene encodes a ligand for KLRK1/NKG2D (natural killer cell lec-tin-like receptor gene 2D) receptor. It consists of α1 and α2 domains, which are connected to the cell surface through glycosylphosphatidylinositol.This receptor is expressed on natural killer (NK) cells, NKT cells, γδ+ T cells and CD8+ T cells.

Immunogen

NKG2D ligand 1 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-ULBP1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

ULBP1 (Unique Long 16 binding protein 1) functions as a stimulatory immune receptor and is expressed on natural killer (NK) cells, NKT cells, γδ+ T cells and CD8+ T cells. The expression of this ligand is stimulated upon infection and other inducers of cellular stress. This ligand binds to the receptor and initiates an immune response by activating multiple signaling pathways in primary NK cells. It serves as a prognostic marker in cervical cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71802

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esther M de Kruijf et al.
BMC cancer, 12, 24-24 (2012-01-20)
Cell surface NKG2D ligands (NKG2DL) bind to the activating NKG2D receptor present on NK cells and subsets of T cells, thus playing a role in initiating an immune response. We examined tumor expression and prognostic effect of NKG2DL in breast
Ryoji Kamei et al.
Oncology letters, 15(1), 747-754 (2018-02-03)
UL16 binding protein 1 (ULBP1) expressed on the tumor cell surface binds to the natural killer group 2 member D (NKG2D) receptor presenting on natural killer (NK), cluster of differentiation (CD)8+ T, and γ δ T cells. However, the roles
Claire Dunn et al.
The Journal of experimental medicine, 197(11), 1427-1439 (2003-06-05)
The activating receptor, NKG2D, is expressed on a variety of immune effector cells and recognizes divergent families of major histocompatibility complex (MHC) class I-related ligands, including the MIC and ULBP proteins. Infection, stress, or transformation can induce NKG2D ligand expression
Hanbyoul Cho et al.
BMC cancer, 14, 957-957 (2014-12-17)
NKG2D (natural killer group 2, member D) is thought to play an important role in mediating the activation of anticancer immune response. Expression of NKG2D ligands (NKG2DLs) is pronounced in malignancies and the heterogeneity of NKG2DL expression remains unclear. Here
Claire L Sutherland et al.
Journal of immunology (Baltimore, Md. : 1950), 168(2), 671-679 (2002-01-05)
The UL16-binding proteins (ULBPs) are a novel family of MHC class I-related molecules that were identified as targets of the human CMV glycoprotein, UL16. We have previously shown that ULBP expression renders a relatively resistant target cell sensitive to NK

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service