Skip to Content
Merck
All Photos(7)

Documents

HPA005480

Sigma-Aldrich

Anti-SYVN1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DER3, Anti-HRD1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

PAPGFPFPPPWMGMPLPPPFAFPPMPVPPAGFAGLTPEELRALEGHERQHLEARLQSLRNIHTLLDAAMLQINQYLTVLASLGPPRPATSVNSTEETATTVVAAASSTSIPSSEATTPTPGASPPAPEMERPPAPESVGT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SYVN1(84447)

General description

Synovial apoptosis inhibitor 1/ synoviolin (SYVN1) is a part of ER-associated degradation (ERAD) and is an E3 ubiquitin ligase. It localizes to endoplasmic reticulum (ER) and is ubiquitously expressed. It is highly expressed in skeletal muscle, pancreas and liver. SYVN1 contains six domains, which span the plasma membrane. It is a non-glycosylated protein and has a cytoplasmic RING-H2 finger domain. This gene is located on chromosome 11q13.

Immunogen

E3 ubiquitin-protein ligase synoviolin precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-SYVN1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Synovial apoptosis inhibitor 1/ synoviolin (SYVN1) plays a key role in various processes, such as embryogenesis or negative regulation of tumor suppressor gene p53. The expression of SYVN1 is induced by IRE1 and ATF6, during ER stress, where it protects against ER-stress induced cell apoptosis. It is a part of endoplasmic reticulum associated degradation (ERAD) and degrades 3-hydroxy-3-methylglutaryl-coenzyme A, along with TCR-α and CD3-δ, which are ERAD substrates. It also sequesters p53, in the cytoplasm, and ubiquitinates it. Thus, it represses the various activities of p53, such as cell cycle regulation and apoptosis. SYVN1 is responsible for maintaining joint homeostasis and hence, is involved in the pathogenesis of arthropathy. In rheumatoid arthritis, it is overexpressed in synovial cells, leading to a ′hyper-ERAD′ state. It interacts with, and degrades tau and phosphorylated tau (p-tau). Therefore, it prevents tau-induced cytotoxicity and helps neuronal survival.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70751

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keisuke Yamamoto et al.
Journal of biochemistry, 144(4), 477-486 (2008-07-31)
Quality control of proteins in the endoplasmic reticulum (ER) is achieved by two mechanisms, the productive folding mechanism, which is assisted by a number of ER-localized molecular chaperones and folding enzymes (collectively termed ER chaperones), and the ER-associated degradation (ERAD)
Masayuki Kaneko et al.
FEBS letters, 581(28), 5355-5360 (2007-10-31)
Human HRD1 and SEL1 are components of endoplasmic reticulum-associated degradation (ERAD), which is a retrograde transport mechanism from the ER to the cytosol for removing unfolded proteins. The expression of HRD1 and SEL1 was induced by ER stress-inducing agents and
Masayuki Kaneko et al.
FEBS letters, 532(1-2), 147-152 (2002-12-03)
Stresses that impair the function of the endoplasmic reticulum (ER) lead to an accumulation of unfolded protein in the ER. Under these conditions, the expression of a variety of genes involved in preventing the accumulation of the unfolded proteins is
Naoko Yagishita et al.
Nature clinical practice. Rheumatology, 4(2), 91-97 (2008-02-01)
Rheumatoid arthritis is a disease associated with painful joints that affects approximately 1% of the population worldwide, and for which no specific cure is available. Among other functions, the endoplasmic reticulum (ER) has an important role in protein folding. When
Y X Shen et al.
Current molecular medicine, 12(2), 138-152 (2012-01-28)
Intraneuronal accumulation of abnormal phosphorylated tau (p-tau) is a molecular pathology in many neurodegenerative tauopathies, including Alzheimer's disease (AD) and frontotemporal dementia with parkinsonism-linked to chromosome 17 (FTDP-17). However, the underlying mechanism remains unclear. Here, we showed an inverse relationship

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service