Skip to Content
Merck
All Photos(1)

Key Documents

H2024

Supelco

N-Hydroxy-3,4-methylenedioxyamphetamine hydrochloride

Synonym(s):

N-Hydroxy-MDA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
Pricing and availability is not currently available.

drug control

USDEA Schedule I; Home Office Schedule 1; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A K Valaer et al.
Journal of chromatographic science, 28(9), 482-486 (1990-09-01)
The reversed-phase liquid chromatographic properties of N-hydroxy-3,4-methylenedioxyamphetamine (NOHMDA) were determined on a C8 stationary phase specifically prepared for the separation of basic compounds. NOHMDA and several N-alkyl MDA derivatives displayed excellent peak shape on this stationary phase without the need
U Braun et al.
Journal of pharmaceutical sciences, 69(2), 192-195 (1980-02-01)
The known central nervous system activity of 3,4-methylenedioxyphenylisopropylamine and its N-methyl homolog prompted the synthesis of a series of analogs with substituents on the nitrogen atom. Most of these analogs (R = alkyl, alkenyl, hydroxy, alkoxy, and alkoxyalkyl) were prepared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service