GERPN203T
Hybond®-NX Nylon hybridization membranes
W × L 20 cm × 3 m, pack of 1 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105339
NACRES:
NB.22
Recommended Products
material
neutral nylon membrane
form
solid
packaging
pack of 1 ea
manufacturer/tradename
Cytiva RPN203T
W × L
20 cm × 3 m
color
White
pore size
0.45 μm pore size
Looking for similar products? Visit Product Comparison Guide
Legal Information
Hybond is a registered trademark of Cytiva
Storage Class Code
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service