Skip to Content
Merck
All Photos(1)

Key Documents

G1288

Sigma-Aldrich

β-(1→3,4,6)-Galactosidase, positionally specific

recombinant, expressed in E. coli, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€602.00

€602.00


Please contact Customer Service for Availability


Select a Size

Change View
1 VIAL
€602.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

€602.00


Please contact Customer Service for Availability

recombinant

expressed in E. coli

Quality Level

form

buffered aqueous solution

specific activity

≥70 units/mg protein

packaging

vial of 0.24 unit

shipped in

wet ice

storage temp.

2-8°C

Biochem/physiol Actions

Releases β(1→3)-, β(1→4)-, and β(1→6)-linked galactose from the non-reducing end of complex oligosaccharides.

Unit Definition

One unit will hydrolyze 1 μmole of p-nitrophenyl β-D-galactopyranoside per min at pH 5.0 at 37 °C.

Physical form

Solution in 20 mM Tris-HCl, pH 7.5, 25 mM NaCl

Preparation Note

Produced from a mixture of enzymes from Streptococcus pneumonia and Xanthamonas sp.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William C Hines et al.
Nature communications, 6, 6927-6927 (2015-04-22)
Clinically relevant human culture models are essential for developing effective therapies and exploring the biology and etiology of human cancers. Current breast tumour models, such as those from oncogenically transformed primary breast cells, produce predominantly basal-like properties, whereas the more

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service