G0375
D-(−)-Galactonic acid γ-lactone
≥99%
Synonym(s):
D-(−)-Galactono-1,4-lactone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H10O6
CAS Number:
Molecular Weight:
178.14
Beilstein:
83008
EC Number:
MDL number:
UNSPSC Code:
12352200
Recommended Products
Assay
≥99%
SMILES string
OC[C@@H](O)[C@@H]1OC(=O)[C@H](O)[C@H]1O
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service