Skip to Content
Merck
All Photos(1)

Documents

F7305

Sigma-Aldrich

Fetal Bovine Serum

Mouse Embryonic Stem Cell Screened, suitable for cell culture, sterile-filtered, USDA Approved

Synonym(s):

FBS, FCS, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207

biological source

bovine fetus

sterility

sterile-filtered

quality

USDA Approved

composition

Hemoglobin, ≤25 mg/dL

technique(s)

cell culture | mammalian: suitable

impurities

≤5 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Fetal Bovine Serum (FBS) is a complex mixture with low and high molecular weight biomolecules exhibiting balanced growth-promoting and growth-inhibiting functions. FBS contains molecules, such as growth factors, proteins, vitamins, trace elements, hormones etc. These molecules are important for the growth and maintenance of cells. It is a common supplement for cell culture media.
Most Fetal Bovine Serum (FBS) batches do not support mouse ES cells. Sigma-Aldrich has pre-screened multiple FBS batches to increase your chances of selecting a batch that will perform within your laboratory, saving you time and money.
In addition to our extensive Fetal Bovine Serum performance and quality assays (mycoplasma, sterility, endotoxin, hemoglobin, etc.), we have developed low density, feeder-free assays to assess Fetal Bovine Serum′s support of mouse embryonic stem cells. Only those batches that performed well with mouse ES cells were selected for this product.

Application

FBS derived from clotted blood is the most widely used undefined supplement in eucaryotic, especially mammalian, cell culture. It is typically used with the classical media that were developed in the 1950s and 1960s. These include many popular media such as Minimum Essential Medium (MEM); Dulbecco′s Modified Eagles Media (DME, DMEM); Iscove′s Modified Eagles Medium (IMDM); M199 and Roswell Park Memorial Institute medium, RPMI-1640.

Analysis Note

  • Endotoxin and hemoglobin tested
  • Mouse embryonic stem cell screened
  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimization of chemically defined cell culture media--replacing fetal bovine serum in mammalian in vitro methods.
van der Valk J et al.
Toxicology in vitro, 24, 1053-1053 (2010)
Alternatives to the use of fetal bovine serum: serum-free cell culture.
Gstraunthaler G
ALTEX, 20, 275-275 (2003)
Li Zhou et al.
Genome biology, 9(7), R119-R119 (2008-07-31)
CD8+ NKT-like cells are naturally occurring but rare T cells that express both T cell and natural killer cell markers. These cells may play key roles in establishing tolerance to self-antigens; however, their mechanism of action and molecular profiles are
Ruth M Risueño et al.
PloS one, 3(3), e1747-e1747 (2008-03-06)
How the T cell antigen receptor (TCR) discriminates between molecularly related peptide/Major Histocompatibility Complex (pMHC) ligands and converts this information into different possible signaling outcomes is still not understood. One current model proposes that strong pMHC ligands, but not weak
Silvia Goldoni et al.
The American journal of pathology, 173(3), 844-855 (2008-08-09)
Decorin, a member of the small leucine-rich proteoglycan gene family, down-regulates members of the ErbB receptor tyrosine kinase family and attenuates their signaling, leading to growth inhibition. We investigated the effects of decorin on the growth of ErbB2-overexpressing mammary carcinoma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service