EP022492080
Eppendorf® epT.I.P.S.® standard
volume 0.1-5 mL, L 120 mm, violet, colorless tip, pkg of 500 ea (5 bags × 100 ea)
Synonym(s):
pipette tips
About This Item
Recommended Products
material
colorless tip
polypropylene
violet
sterility
non-sterile
packaging
pkg of 500 ea (5 bags × 100 ea)
manufacturer/tradename
Eppendorf® 022492080
L
120 mm
volume
0.1-5 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Original, high-quality Eppendorf pipette tips packaged in resealable bags
- Available in sizes from 10 µL to 10 mL in Eppendorf Quality purity grade
- epT.I.P.S. Standard 200 µL, 300 µL and 1,000 µL tips are also available color-coded yellow and blue
- epT.I.P.S. Standard pipette tips can be autoclaved at 121 °C in 20 min.
- Fine retracted or extended tips simplify work with small volumes
Features and Benefits
- Original, high-quality Eppendorf pipette tips packaged in resealable bags
- Available in sizes from 10 µL to 10 mL in Eppendorf Quality purity grade
- epT.I.P.S. Standard 200 µL, 300 µL and 1,000 µL tips are also available color-coded yellow and blue
- epT.I.P.S. Standard pipette tips can be autoclaved at 121 °C in 20 min.
- Fine retracted or extended tips simplify work with small volumes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service