EP0030077547
Eppendorf® ep Dualfilter T.I.P.S.®
10 racks x 96 ea, PCR clean, capacity 2-100 μL, sterile
Synonym(s):
pipetting tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
Recommended Products
material
yellow (Tray)
sterility
sterile
feature
DNase free
RNase free
filter
packaging
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Eppendorf® 0030077547
technique(s)
PCR: suitable
capacity
2-100 μL
maximum volume
100 μL
Looking for similar products? Visit Product Comparison Guide
General description
- Eppendorf® ep Dualfilter T.I.P.S.® of 2-100 μL capacity are well suited for applications with samples that contain detergents.
- Two filter layers provide dual protection against aerosol and biomolecules to guard your pipette and sample.
- Available as PCR clean/Sterile (sterile and pyrogen-free).
- Continuous inspection of each production lot by an independent laboratory guarantees high purity and quality.
- Ultrahomogenous surface for maximum reproducibility.
- Ultrahydrophobic surface for minimal sample loss.
- Significantly reduced foam formation during pipetting.
- Extreme resistance to chemicals.
- Available in PCR clean and Eppendorf Quality.
- Also available as ep Dualfilter T.I.P.S. in PCR clean/Sterile.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
ep Dualfilter T.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service