Skip to Content
Merck
All Photos(1)

Key Documents

D7033

Sigma-Aldrich

DL-threo-Dihydrosphingosine

≥98%

Synonym(s):

DL-threo-1,3-Dihydroxy-2-aminooctadecane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H39NO2
CAS Number:
Molecular Weight:
301.51
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥98%

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCC[C@@](O)([H])[C@@]([H])(N)CO

InChI

1S/C18H39NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(21)17(19)16-20/h17-18,20-21H,2-16,19H2,1H3/t17-,18-/m0/s1

InChI key

OTKJDMGTUTTYMP-ROUUACIJSA-N

Biochem/physiol Actions

Sphingosine kinase inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Tokuda et al.
Journal of cellular biochemistry, 72(2), 262-268 (1999-02-18)
In osteoblast-like MC3T3-E1 cells, we have recently reported that sphingosine 1-phosphate among sphingomyelin metabolites acts as a second messenger for tumor necrosis factor-alpha (TNF)-induced interleukin-6 (IL-6) synthesis. In the present study, we investigated the effect of extracellular sphingomyelinase on IL-6
G S Dbaibo et al.
Proceedings of the National Academy of Sciences of the United States of America, 92(5), 1347-1351 (1995-02-28)
Ceramide, a lipid mediator, has been most closely associated with antiproliferative activities. In this study, we examine the mechanism by which ceramide induces growth suppression and the role of the retinoblastoma gene product (Rb) in this process. Withdrawal of serum
O Kozawa et al.
Journal of cellular biochemistry, 70(3), 338-345 (1998-08-26)
We previously reported that prostaglandin (PG)E1 and PGF2alpha induce the synthesis of interleukin-6 (IL-6) via activation of protein kinase (PK)A and PKC, respectively, in osteoblast-like MC3T3-E1 cells. In addition, we have shown that basic fibroblast growth factor (bFGF) elicits IL-6

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service