D2256
Dextrin from corn
commercial grade, Type III, powder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
Recommended Products
grade
commercial grade
type
Type III
form
powder
color
yellow
SMILES string
OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O
InChI
1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1
InChI key
WQZGKKKJIJFFOK-DVKNGEFBSA-N
Gene Information
human ... PYGM(5837)
Looking for similar products? Visit Product Comparison Guide
Analysis Note
Approx. 75% water-soluble.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Glucose polymer supplements in low birth weight infants.
O G Brooke
British medical journal (Clinical research ed.), 286(6371), 1143-1143 (1983-04-02)
Toshikazu Sugimoto et al.
Journal of industrial microbiology & biotechnology, 38(12), 1985-1991 (2011-05-28)
Submerged batch cultures of Aspergillus kawachii grown on indigestible dextrin were investigated for potential improvements in glucoamylase (GA) production. In flask culture, specific GA productivities per dry weight biomass using dextrin and indigestible dextrin were 11.0 and 56.1 mU/mg-DW, respectively. Indigestible
O E Odeleye et al.
European journal of clinical nutrition, 46(5), 367-371 (1992-05-01)
The effect of caloreen (glucose polymer) supplementation on indicators of iron status during protein-energy malnutrition was studied. Sixty-four children with moderate protein energy malnutrition (PEM) were fed diets supplemented with caloreen or starch (control) for 14 days, following which iron
Galina A Kovalenko et al.
Carbohydrate research, 343(7), 1202-1211 (2008-03-19)
Glucoamylase (GA) was immobilized by adsorption on carbon support: on Sibunit, on bulk catalytic filamentous carbon (bulk CFC) and on activated carbon (AC). This was used to prepare heterogeneous biocatalysts for the hydrolysis of starch dextrin. The effect of the
Glucose-polymer in acute diarrhoea.
Archives of disease in childhood, 57(9), 721-722 (1982-09-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service