CLS6555
Corning® multiwell plates, plate lids and sealing mats
Thermowell 96 well plate sealing mat, silicone rubber sealing mat, non-sterile, cs/25
Synonym(s):
96 well plate sealing mat, silicone rubber sealing mat
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41100000
Recommended Products
material
silicone
sterility
non-sterile
feature
lid
packaging
pack of 1
case of 25
manufacturer/tradename
Corning 6555
size
96 wells
color
none
Looking for similar products? Visit Product Comparison Guide
General description
- Thermowell sealing mats for 96 well PCR plates are easy to apply and remove
- Fully autoclavable and reusable (at least five times)
- These silicone rubber mats offer a cost effective alternative to other sealing methods and provide 100% sealing when used in conjunction with clamp or screw-down heated lid thermal cyclers
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service