CLS430776
Corning® large volume centrifuge tubes
250 mL centrifuge tubes, polypropylene, w/ plug seal cap, sterile, 102/cs
Synonym(s):
250 ml centrifuge tubes, centrifuge tubes, large volume centrifuge tubes
About This Item
Recommended Products
material
cap (plugseal)
conical bottom tube
polypropylene
sterility
sterile (non-pyrogenic)
feature
RCF 6000 × g
disposable
graduations 1.5 ‑ 14.5 mL
packaging
case of 102
pack of 6 ×
manufacturer/tradename
Corning 430776
parameter
6000 × g max. RCF
cap diam.
1.33 in.
capacity
250 mL
tube O.D.
2.35 in.
color
natural
fitting
cap (Plug Seal)
suitability
suitable for (applications requiring large volume centrifugation)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Support cushions (Catalog No. CLS430236 for 250mL centrifuge tubes) must be used with this product unless the centrifuge rotor has appropriate V-bottom cushions
- Polyethylene plug seal caps
- Maximum RCF of 6,000xg
- Tubes are sterile and certified nonpyrogenic
Support cushions must be used with this product unless the centrifuge rotor has V-bottom holders.
Legal Information
recommended
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service