Skip to Content
Merck
All Photos(1)

Documents

CLS3969

Corning® CellSTACK® accessories

screw cap, not vented

Synonym(s):

Corning cellstack cell culture chambers, tissue culture chambers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122104
NACRES:
NB.13

material

high-density polyethylene

sterility

sterile

feature

disposable

packaging

case of 6

manufacturer/tradename

Corning 3969

color

black

Looking for similar products? Visit Product Comparison Guide

General description

  • 33 mm threaded cap
  • Solid cap is used in closed systems or applications where venting is not necessary
  • Sterilized by gamma irradiation
Available for filling, venting, stacking and manipulation.

Legal Information

CellSTACK is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service