Skip to Content
Merck
All Photos(1)

Documents

L-41(HeLa derivative)

96121716, human cervix, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

L-41(HeLa derivative), 96121716

biological source

human cervix

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human cervix carcinoma

Cell Line Description

The cell line L41 was reported to have been established by cloning the line J96, which was originally derived from the bone marrow of a child patient with leukaemia. However, it has been found to be indistinguishable from Hela by STR PCR DNA profiling.Therefore, the cell line should be considered as being derived from HeLa. The cell line L41 was mycoplasma eradicated at ECACC. Ethnicity: Black.

Culture Medium

RPMI 1640 or DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:3 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service