Skip to Content
Merck
All Photos(1)

Key Documents

NFS-25 C-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

mouse blood

packaging

tube of 5 μg 93090713-DNA-5UG
pkg of vial of cells 93090713-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Not specified

receptors

Ly-17, Lyb-2, Ly-1, Ly-5 (B220)

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse pre-B lymphoblast

Cell Line Description

Derived from a spontaneous lymphoma in an NFS.C58v-1 mouse. These pre-B cells are negative for surface Ig, Ia and ThB.

Culture Medium

DMEM + 10% NCTC 109 + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 1% Sodium Pyruvate (NaP) + 1mM Oxaloacetic acid + 50μM 2-Mercaptoethanol (2ME) + 2μg/ml Transferrin + 0.2IU/ml Insulin + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Immediately after resuscitation a pre-centrifugation step to remove cryoprotectant (DMSO) is recommended i.e. pellet the cells by centrifugation at 100 x g for 5 minutes and resuspend the cell

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service