Skip to Content
Merck
All Photos(1)

Documents

HL60 Ast.4

88120802, human blood, Promyeloid, blast/promyelocyte

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

HL60 Ast.4, 88120802

biological source

human blood

growth mode

Suspension

karyotype

2n = 48, hyperdiploid

morphology

Promyeloid, blast/promyelocyte

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human Caucasian promyelocytic leukaemia

Cell Line Description

Variant of the promyelocytic cell line HL60 isolated after treating cells with α-particle radiation and then cloned in medium containing 1.25% DMSO. Treatment with 50nM TPA results in only very limited differentiation towards basophilic morphology (Giemsa stain). Differentiation towards monocyte lineage has not been observed.

Application

Differentiation studies

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) + 1.25% Dimethyl Sulphoxide (DMSO).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Do not allow the culture medium to become acidic.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service