Skip to Content
Merck
All Photos(1)

Documents

RAJI Cell Line human

85011429, human lymph, Lymphoblastoid

Synonym(s):

GM04671 Cells, P1-Raji Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

RAJI Cell Line human, 85011429

biological source

human lymph

description

Human Black, Burkitt′s lymphoma

growth mode

Suspension

karyotype

2n = 46, diploid

morphology

Lymphoblastoid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human Burkitt′s lymphoma

Cell Line Description

Established in 1963 from Burkitt′s lymphoma in an 11 year old male. Growth is in the form of single cells without attachment and as macroscopically visible clumps containing many hundreds of cells. Resistant to VSV. This cell line carries the latent Epstein-Barr Virus (EBV) genome and is positive for EBNA. RAJI is sometimes referred to as a ′non-producer′; the EBV genome carries deletions attributed to preventing the formation of virus particles. Ethnicity: Black.

Application

Resistance and virus studies: vesicular stomatitis virus (VSV).
RAJI Cell Line human has been used to investigate the transport of V. parahaemolyticus across the Payer′s patch of the intestinal epithelium and it has been co-cultured with Caco-2 (human colorectal adenocarcinoma cell-line) monolayers to study bacterial translocation.

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,12
D13S317: 13
D16S539: 8,11
D5S818: 10,13
D7S820: 10
THO1: 6,7
TPOX: 8,13
vWA: 16,19

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service