C9811
CryoCane™ aluminum canes
capacity, 5 vial(s)
Synonym(s):
cryogenic vial cane, cryostorage cane, cryovial cane, dewar cane, liquid nitrogen cane
All Photos(1)
About This Item
Recommended Products
feature
capacity 5 vial(s)
manufacturer/tradename
Nalgene®
Overall L
290 mm
Related Categories
General description
Firmly holds 1.2 or 2.0 mL Nalgene cryogenic vials for storage in Dewar-type, liquid nitrogen freezers.
Projections support base of each vial.
Legal Information
CryoCane is a trademark of Thermo Fisher Scientific or its subsidiaries
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries