Skip to Content
Merck
All Photos(1)

Key Documents

C8610

Sigma-Aldrich

Casarett and Doull′s Toxicology: The Basic Science of Poisons, 6th ed.

Sign Into View Organizational & Contract Pricing


About This Item

ISBN-10:
0-07-134721-6
ISBN-13:
978-0-07-134721-1
UNSPSC Code:
55101509

publication info

C. Klaasen, McGraw-Hill, 2001, 1275 pp., hard cover

manufacturer/tradename

Mcgraw-Hill

General description

Long-established as the gold standard in the field, this sixth edition equips one with an unsurpassed treatment of modern toxicology, including the principles, concepts, mechanisms, and modes of thought that are the foundation of the discipline. This classic book includes a full discussion of toxicokinetics; how each body system responds to poisons; and the specific effects of a wide range of toxic agents pesticides to radiation. It covers recent advances in molecular biology, pharmacogenetics, clinical toxicology, and behavioral/clinical research.

Table of Contents

General Principles of Toxicology.
Disposition of Toxicants.
Non-Organ Directed Toxicity.
Target Organ Toxicity.
Toxic Agents.
Environmental Toxicology.
Applications of Toxicology.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service