Skip to Content
Merck
All Photos(1)

Documents

B9304

Sigma-Aldrich

Anti-Bcl-x antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-BCL2L1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 26-30 kDa

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:1,000 using human colon adenocarcinoma sections
microarray: suitable
western blot: 1:2,000 using human Burkit lymphoma Raji cell lysate

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BCL2L1(598)
mouse ... Bcl2l1(12048)
rat ... Bcl2l1(24888)

General description

Bcl-2-like protein 1 (BCL2L1) or Bcl-2-related gene x, long isoform (Bcl-x) gene is mapped to human chromosome 20q11.21. BCL2L1 belongs to the Bcl-2 family. The bcl-x gene on alternative splicing leads two distinct proteins. It comprises multiple Bcl-2 homology (BH3) domains. Bcl-xL is expressed in the adult brain.(10)

Specificity

The antibody reacts with Bcl-xL.

Immunogen

synthetic peptide corresponding to the N-terminal of human Bcl-x (amino acids 2-20) conjugated to BSA. This sequence is identical in human and mouse Bcl-xS, Bcl-xL, Bcl-Xβ and Bcl-Xγ isoforms and is highly conserved in rat Bcl-xS and Bcl-xL (single amino acid substitution).

Application

Anti-Bcl-x antibody produced in rabbit may be used in immunoblotting and immunohistochemistry.

Biochem/physiol Actions

Bcl-2-like protein 1 (BCL2L1) or Bcl-2-related gene x arelong isoforms (Bcl-x) of proteins that are involved in regulating apoptosis. Similar to Bcl-2, Bcl-xL can inhibit cell death upon growth factor removal, when transfected in an IL-3 dependent cell line. Bcl-xL can form ion channels in biological membranes and may maintain cell survival by regulating the permeability of intracellular membranes. Bcl-xL may bind to apoptotic protease activating factor 1 (Apaf-1) preventing it from activating caspase 9. Like Bcl-2, Bcl-x activity is regulated by specific phosphorylation of a 60-amino acid regulatory loop region. Bcl-xL has been shown to regulate cell response to oxidative stress, cellular resistance to DNA damage-induced apoptosis by chemotherapeutic agents and ionizing radiation, and favors neurite outgrowth.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month.For extended storage, freeze in working aliquots.Repeated freezing and thawing is not recommended.Storage in "frost-free" freezers is not recommended. Ifslight turbidity occurs upon prolonged storage, clarifythe solution by centrifugation before use. Workingdilution samples should be discarded if not used within12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Megan Stevens et al.
Frontiers in genetics, 10, 804-804 (2019-09-26)
Apoptosis plays a vital role in cell homeostasis during development and disease. Bcl-x, a member of the Bcl-2 family of proteins, is a mitochondrial transmembrane protein that functions to regulate the intrinsic apoptosis pathway. An alternative splicing (AS) event in
A J Minn et al.
Nature, 385(6614), 353-357 (1997-01-23)
Bcl-2-related proteins are critical regulators of cell survival that are localized to the outer mitochondrial, outer nuclear and endoplasmic reticulum membranes. Despite their physiological importance, the biochemical function of Bcl-2-related proteins has remained elusive. The three-dimensional structure of Bcl-xL, an
Chloe F A Warren et al.
Cell death & disease, 10(3), 177-177 (2019-02-23)
The BCl-2 family has long been identified for its role in apoptosis. Following the initial discovery of BCL-2 in the context of B-cell lymphoma in the 1980s, a number of homologous proteins have since been identified. The members of the
Jianfang Wang et al.
Cellular signalling, 23(12), 2030-2038 (2011-08-16)
Functional analysis of a Bcl-xL phosphorylation mutant series has revealed that cells expressing Bcl-xL(Ser49Ala) mutant are less stable at G2 checkpoint after DNA damage and enter cytokinesis more slowly after microtubule poisoning, than cells expressing wild-type Bcl-xL. These effects of
Katherine Amps et al.
Nature biotechnology, 29(12), 1132-1144 (2011-11-29)
The International Stem Cell Initiative analyzed 125 human embryonic stem (ES) cell lines and 11 induced pluripotent stem (iPS) cell lines, from 38 laboratories worldwide, for genetic changes occurring during culture. Most lines were analyzed at an early and late

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service