Skip to Content
Merck
All Photos(1)

Key Documents

B5014

Sigma-Aldrich

Met-Lys-Bradykinin acetate salt

≥97% (HPLC)

Synonym(s):

Met-Kallidin, Met-Lys-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M S Araujo-Viel et al.
Hoppe-Seyler's Zeitschrift fur physiologische Chemie, 362(3), 337-345 (1981-03-01)
The synthetic tridecapeptide Gly-Leu-Met-Lys-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg was used as a model substrate for horse urinary and porcine pancreatic kallikreins. The Met-Lys bond is hydrolyzed selectively by both enzymes. Oxidation of the methionine residue to sulfoxide made the peptide resistant to both kallikreins.
Elliot, D.F. and Lewis, G.P.
Journal of Biochemistry, 95, 437-437 (1965)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service