Skip to Content
Merck
All Photos(1)

Key Documents

AV51761

Sigma-Aldrich

Anti-CYP1B1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CP1B, Anti-Cytochrome P450, family 1, subfamily B, polypeptide 1, Anti-GLC3A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

61 kDa

species reactivity

mouse, guinea pig, horse, human, canine, rat, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CYP1B1(1545)

Immunogen

The immunogen for anti-CYP1B1 antibody: synthetic peptide derected towards the middle region of human CYP1B1

Application

Anti-CYP1B1 antibody produced in rabbit is suitable for western blotting at a concentration of 0.5 μg/ml.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Cytochrome P450, family 1, subfamily B, polypeptide 1 (CYP1B1) is a member of cytochrome P450 superfamily of monooxygenases. CYP1B1 is localized to the endoplasmic reticulum.[1] It metabolizes polycyclic aromatic hydrocarbon and 17β-estradiol[1] and is possibly involved in the metabolism of signaling molecule involved in the development of the eye.[2] Mutations in CYP1B1 are associated with primary congenital glaucoma[2][3] and congenital anterior staphylomas.[4]

Sequence

Synthetic peptide located within the following region: AVANVMSAVCFGCRYSHDDPEFRELLSHNEEFGRTVGAGSLVDVMPWLQY

Physical form

Lyophilized from PBS buffer with 2% sucrose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xueli Chen et al.
The British journal of ophthalmology, 98(2), 246-251 (2013-11-15)
The aim of the present work was to investigate CYP1B1 gene mutations in patients of Han Chinese ethnicity with primary congenital glaucoma (PCG), and explore the clinical characteristics associated with operative effects. Peripheral blood genomic DNA was extracted from patients
Milana Kokosar et al.
Scientific reports, 6, 22883-22883 (2016-03-16)
Genetic and epigenetic factors may predispose women to polycystic ovary syndrome (PCOS), a common heritable disorder of unclear etiology. Here we investigated differences in genome-wide gene expression and DNA methylation in adipose tissue from 64 women with PCOS and 30
Ramzi Al Judaibi et al.
Clinical ophthalmology (Auckland, N.Z.), 8, 445-448 (2014-03-05)
Here, we present two patients with congenital anterior staphyloma, with mutations in the CYP1B1 gene. We reviewed the medical records, including the genetic analysis. Two unrelated patients presented with congenital anterior staphylomas. Both patients showed mutations in the CYP1B1 gene.
Douglas B Gould et al.
The International journal of developmental biology, 48(8-9), 1015-1029 (2004-11-24)
Development of the ocular anterior segment involves a series of inductive interactions between neural ectoderm, surface ectoderm and periocular mesenchyme. The timing of these events is well established but less is known about the molecular mechanisms involved. Various genes that
The nuclear vitamin D receptor: biological and molecular regulatory properties revealed.
M R Haussler et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 13(3), 325-349 (1998-04-03)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service