Skip to Content
Merck
All Photos(1)

Key Documents

AV32872

Sigma-Aldrich

Anti-ZBTB7C antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Zinc finger and BTB domain containing 7C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

69 kDa

species reactivity

rat, horse, guinea pig, dog, rabbit, goat, human, mouse, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZBTB7C(201501)

Related Categories

General description

ZBTB7C is a zinc-finger protein that functions as a tumor suppressor. Decreased ZBTB7C expression has been linked to cervical carcinoma.
Rabbit Anti-ZBTB7C antibody recognizes rat, mouse, bovine, human, chicken, and canine ZBTB7C.

Immunogen

Synthetic peptide directed towards the N terminal region of human ZBTB7C

Application

Rabbit Anti-ZBTB7C antibody can be used for western blotting applications at a concentration of 5μg/ml.

Biochem/physiol Actions

The function of Anti-ZBTB7C has not yet been determined.

Sequence

Synthetic peptide located within the following region: MANDIDELIGIPFPNHSSEVLCSLNEQRHDGLLCDVLLVVQEQEYRTHRS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xueying An et al.
Biomedical journal, 47(3), 100651-100651 (2023-08-11)
Dysregulation of long non-coding RNAs (lncRNAs) is an important component of tumorigenesis. Aberrant expression of lncRNA taurine upregulated gene 1 (lncTUG1) has been reported in various tumors; however, its precise role and key targets critically involved in osteosarcoma (OS) progression
Martina Schmitz et al.
PloS one, 7(6), e39632-e39632 (2012-07-05)
HPV DNA integration into the host genome is a characteristic but not an exclusive step during cervical carcinogenesis. It is still a matter of debate whether viral integration contributes to the transformation process beyond ensuring the constitutive expression of the
Johanna Sandgren et al.
Experimental & molecular medicine, 42(7), 484-502 (2010-06-11)
Epigenomic and genomic changes affect gene expression and contribute to tumor development. The histone modifications trimethylated histone H3 lysine 4 (H3K4me3) and lysine 27 (H3K27me3) are epigenetic regulators associated to active and silenced genes, respectively and alterations of these modifications

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service