Skip to Content
Merck
All Photos(2)

Documents

AV32331

Sigma-Aldrich

Anti-EYA3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Eyes absent homolog 3 (Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

63 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EYA3(2140)

General description

EYA3 interacts with Six1 and subsequently activates TSHβ expression in response to light changes in the photoperiodic system of mice. EYA3 expression has also been implicated in Ewing sarcoma.
Rabbit Anti-EYA3 antibody recognizes human, mouse, rat, and canine EYA3.

Immunogen

Synthetic peptide directed towards the middle region of human EYA3

Application

Rabbit Anti-EYA3 antibody can be used for IHC (4-8μg/ml) and western blot (0.05-1.0μg/ml) applications.

Biochem/physiol Actions

Eyes absent homolog 3 (EYA3) is a member of the eyes absent (EYA) family of proteins. This protein may act as a transcriptional activator and have a role during development. A similar protein in mice can act as a transcriptional activator. Two transcript variants encoding distinct isoforms have been identified for this gene.

Sequence

Synthetic peptide located within the following region: QSRKNMTSKNRGKRKADATSSQDSELERVFLWDLDETIIIFHSLLTGSYA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tyler P Robin et al.
Molecular cancer research : MCR, 10(8), 1098-1108 (2012-06-23)
Ewing sarcoma is an aggressive pediatric cancer of the bone and soft tissue, in which patients whose tumors have a poor histologic response to initial chemotherapy have a poor overall prognosis. Therefore, it is important to identify molecules involved in
Koh-Hei Masumoto et al.
Current biology : CB, 20(24), 2199-2206 (2010-12-07)
Living organisms detect seasonal changes in day length (photoperiod) [1-3] and alter their physiological functions accordingly to fit seasonal environmental changes. TSHβ, induced in the pars tuberalis (PT), plays a key role in the pathway that regulates vertebrate photoperiodism [4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service