form
solid
mol wt
monoisotopic mol wt 1114.57 Da
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... HIST1H1A(3024) , HIST1H1C(3006) , HIST1H1D(3007) , HIST1H1E(3008) , HIST1H1T(3010)
mouse ... Hist1h1c(50708) , Hist1h1d(14957) , Hist1h1e(50709) , Hist1h1t(107970)
rat ... H1f4(201097) , Hist1h1t(24438) , LOC684681(684681) , LOC684828(684828)
Application
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service