Skip to Content
Merck
All Photos(1)

Key Documents

A4440

Sigma-Aldrich

Asp-Asp-Asp-Asp

≥97% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22N4O13
CAS Number:
Molecular Weight:
478.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (TLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

NC(CC(O)=O)C(=O)NC(CC(O)=O)C(=O)NC(CC(O)=O)C(=O)NC(CC(O)=O)C(O)=O

InChI

1S/C16H22N4O13/c17-5(1-9(21)22)13(29)18-6(2-10(23)24)14(30)19-7(3-11(25)26)15(31)20-8(16(32)33)4-12(27)28/h5-8H,1-4,17H2,(H,18,29)(H,19,30)(H,20,31)(H,21,22)(H,23,24)(H,25,26)(H,27,28)(H,32,33)

InChI key

OZRFYUJEXYKQDV-UHFFFAOYSA-N

Amino Acid Sequence

Asp-Asp-Asp-Asp

Biochem/physiol Actions

Tetraaspartic acid (Asp-Asp-Asp-Asp) is a small polyanionic peptide used in physicochemical studies along with other polyanionic peptides. Tetraaspartic acid forms complexes with metals such as copper making it useful in studies on copper mediated effects such as free radial formation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xiaofei Li et al.
Biotechnology for biofuels, 6(1), 94-94 (2013-07-04)
Plant cell wall polysaccharide composition varies substantially between species, organs and genotypes. Knowledge of the structure and composition of these polysaccharides, accompanied by a suite of well characterised glycosyl hydrolases will be important for the success of lignocellulosic biofuels. Current
Csilla Kállay et al.
Journal of inorganic biochemistry, 99(7), 1514-1525 (2005-06-02)
Copper(II) complexes of di-, tri- and tetra-peptides built up from Asp and/or Glu residues were studied by potentiometric and various spectroscopic techniques including UV-visible, circular dichroism and electron paramagnetic resonance measurements. The ligands contain two to five carboxylate functions and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service