Skip to Content
Merck
All Photos(1)

Key Documents

A2722

Sigma-Aldrich

[Val671]-Amyloid β/A4 Precursor Protein 770 Fragment 667-676 trifluoroacetate salt

≥95% (HPLC), solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H82N14O18
CAS Number:
Molecular Weight:
1179.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@@H](N)CO)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](C)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](Cc1ccccc1)C(=O)N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C51H82N14O18/c1-25(2)39(65-45(77)32(17-19-37(69)70)59-42(74)29(53)24-66)48(80)60-30(14-9-10-20-52)44(76)64-40(26(3)4)49(81)63-35(23-38(71)72)46(78)57-27(5)41(73)58-31(16-18-36(67)68)43(75)62-34(22-28-12-7-6-8-13-28)47(79)61-33(50(82)83)15-11-21-56-51(54)55/h6-8,12-13,25-27,29-35,39-40,66H,9-11,14-24,52-53H2,1-5H3,(H,57,78)(H,58,73)(H,59,74)(H,60,80)(H,61,79)(H,62,75)(H,63,81)(H,64,76)(H,65,77)(H,67,68)(H,69,70)(H,71,72)(H,82,83)(H4,54,55,56)/t27-,29-,30-,31-,32-,33-,34-,35-,39-,40-/m0/s1

InChI key

XVRVLVNUXPHGIL-ZSDPNEGOSA-N

Gene Information

human ... APP(351)

Amino Acid Sequence

Ser-Glu-Val-Lys-Val-Asp-Ala-Glu-Phe-Arg

Biochem/physiol Actions

β-secretase substrate corresponding to the Met/Val substitution of the amyloid precursor protein (APP) β-secretase cleavage site.
β-secretase substrate; corresponds to Met/Val substitution of the amyloid precursor protein (APP) β-secretase cleavage site.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service