A2177
Arphamenine B hemisulfate salt hydrate
from microbial
Synonym(s):
(2R,5S)-5-Amino-8-guanido-4-oxo-2-p-hydroxyphenylmethyloctanoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C16H24N4O4 · 1/2H2SO4
CAS Number:
Molecular Weight:
385.43
MDL number:
UNSPSC Code:
12352204
biological source
microbial
storage temp.
−20°C
SMILES string
OS(O)(=O)=O.NC(CCCNC(N)=N)C(=O)CC(Cc1ccc(O)cc1)C(O)=O.NC(CCCNC(N)=N)C(=O)CC(Cc2ccc(O)cc2)C(O)=O
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Arphamenines A and B, new inhibitors of aminopeptidase B, produced by bacteria.
H Umezawa et al.
The Journal of antibiotics, 36(11), 1572-1575 (1983-11-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service