Skip to Content
Merck
All Photos(1)

Key Documents

8

Sigma-Aldrich

Sodium 2-ethylhexyl sulfate

Type 8, ~40%

Synonym(s):

2-Ethylhexyl sulfate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17NaO4S
CAS Number:
Molecular Weight:
232.27
Beilstein:
5177087
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

type

Type 8

concentration

~40%

density

1.12 g/mL at 20 °C (lit.)

SMILES string

[Na+].CCCCC(CC)COS([O-])(=O)=O

InChI

1S/C8H18O4S.Na/c1-3-5-6-8(4-2)7-12-13(9,10)11;/h8H,3-7H2,1-2H3,(H,9,10,11);/q;+1/p-1

InChI key

DGSDBJMBHCQYGN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Alkyl sodium sulfate surfactants.

Linkage

These are the same anionic surfactants formerly produced by Union Carbide under the Tergitol name.

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W M Kluwe
Environmental health perspectives, 65, 271-278 (1986-03-01)
Chronic toxicity and carcinogenicity studies of several phthalic acid esters (PAEs) and compounds containing a 2-ethylhexyl moiety were conducted in Fischer 344 rats and B6C3F1 (hybrid) mice. The compounds studied were phthalic anhydride, di(2-ethylhexyl) phthalate, butyl benzyl phthalate, diallyl phthalate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service