Skip to Content
Merck
All Photos(1)

Key Documents

76270

Sigma-Aldrich

Patent Blue V calcium salt

for microscopy

Synonym(s):

Acid Blue 3 calcium salt, Acid Blue V calcium salt, Food Blue 5, Patent Blue V hemicalcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31N2O7S2 · 0.5Ca
CAS Number:
Molecular Weight:
579.71
Colour Index Number:
42051
Beilstein:
4108121
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for microscopy

SMILES string

[Ca++].CCN(CC)c1ccc(cc1)\C(=C2/C=C\C(C=C2)=[N+](\CC)CC)c3cc(O)c(cc3S([O-])(=O)=O)S([O-])(=O)=O.CCN(CC)c4ccc(cc4)C(=C5/C=C\C(C=C5)=[N+](\CC)CC)\c6cc(O)c(cc6S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/2C27H32N2O7S2.Ca/c2*1-5-28(6-2)21-13-9-19(10-14-21)27(20-11-15-22(16-12-20)29(7-3)8-4)23-17-24(30)26(38(34,35)36)18-25(23)37(31,32)33;/h2*9-18H,5-8H2,1-4H3,(H2-,30,31,32,33,34,35,36);/q;;+2/p-2

InChI key

RTMBGDBBDQKNNZ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Other Notes

Literature for Patent Blue Violet discusses two dyes which are very similar, Color Index Numbers 42045 and 42051. Structurally they differ with regard to the substituents on one of the aromatic rings. C.I. 42045 is 2,4-disulfo-substituted while C.I. 42051 is 2,4-disulfo-5-hydroxy-substituted. This is the latter form.

comparable product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service