Skip to Content
Merck
All Photos(1)

Key Documents

75926

Sigma-Aldrich

Oxonol VI

suitable for fluorescence, ≥95% (UV)

Synonym(s):

1,5-Bis(5-oxo-3-propylisoxazol-4-yl)pentamethine oxonol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N2O4
CAS Number:
Molecular Weight:
316.35
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

Assay

≥95% (UV)

form

powder

solubility

methanol: soluble

fluorescence

λex 523 nm; λem 630 nm in 0.1 M Tris pH 9.0

suitability

suitable for fluorescence

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCC1=NOC(=O)\C1=C/C=C/C=C/c2c(O)onc2CCC

InChI

1S/C17H20N2O4/c1-3-8-14-12(16(20)22-18-14)10-6-5-7-11-13-15(9-4-2)19-23-17(13)21/h5-7,10-11,20H,3-4,8-9H2,1-2H3/b7-5+,10-6+,13-11-

InChI key

CPHARWHMAGUXDW-PQWGHOHWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Oxonol VI can be used as a fluorescent potentiometric indicator to detect the membrane potentials in lipid vesicles. Quantitative measurements of the membrane potentials can be easily calibrated using oxonol VI because of its response to membrane potential changes.[1][2] It is also used as an electric potential-sensitive probe to study membrane vesicles from E. coli cells.[3]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

As an optical indicator for membrane potentials in lipid vesicles[4]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ratiometric fluorescence measurements of membrane potential generated by yeast plasma membrane H+-ATPase reconstituted into vesicles
Holoubek A, et al.
Biochimica et Biophysica Acta - Biomembranes, 1609(1), 71-79 (2003)
Preparation of Everted Membrane Vesicles from Escherichia coli Cells
Verkhovskaya M
Bio-protocol, 7(9), e2254-e2254 (2017)
Selective electrodiffusion of zinc ions in a Zrt-, Irt-like protein, ZIPB
Lin W, et al.
Journal of biological and chemical chronicles, 285(50), 39013-39020 (2010)
J Fumero et al.
Biochimica et biophysica acta, 944(2), 164-176 (1988-10-06)
Differential scanning calorimetry (DSC) has been employed to determine the effect of five commonly employed extrinsic potential-sensitive probes on phase transitions of multilamellar suspensions of L-alpha-dimyristoylphosphatidylcholine (DMPC). At mol% values of less than five, the effect of these probes on
R J Clarke et al.
Biochimica et biophysica acta, 981(2), 326-336 (1989-06-06)
A method is described for studying the coupling ratio of the Na+/K+ pump, i.e., the ratio of pump-mediated fluxes of Na+ and K+, in a reconstituted system. The method is based on the comparison of the pump-generated current with the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service