Skip to Content
Merck
All Photos(1)

Key Documents

32856

Sigma-Aldrich

May-Grünwald solution

according to May-Grünwald

Synonym(s):

Eosin – methylene blue solution, Eosin methylene blue according to May-Grünwald

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
Pricing and availability is not currently available.

quality

according to May-Grünwald

suitability

passes test for UV spectrum

SMILES string

[K+].COC(=O)c1ccccc1C2=C3C=C(Br)C(=O)C(Br)=C3Oc4c(Br)c([O-])c(Br)cc24

InChI

1S/C21H10Br4O5.K/c1-29-21(28)9-5-3-2-4-8(9)14-10-6-12(22)17(26)15(24)19(10)30-20-11(14)7-13(23)18(27)16(20)25;/h2-7,26H,1H3;/q;+1/p-1

InChI key

NXBZXYJXNIFCRJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

May-Grünwald-Giemsa (MGG) stain is a Romanowsky-type, polychromatic stain as those of Giemsa, Leishman and Wright. It is used as reference stain in haematology. It is also used as routine stain in diagnostic cytopathology for the study of air-dried preparations (centrifuged body fluids, lymph node imprints, and fine needle aspirations).[1]

Application

May-Grünwald and Giemsa stain was used for staining polymorphonuclear cells, monocytes and lymphocytes, for differential leukocyte counts, during hematological analysis, in an experimental study conducted to study immune responses in the pathology of acute magnesium deficiency in rats.[2] It was also used to stain cells from bronchoalveolar lavage (BAL) fluid, in an experimental investigation, done to study the role of Fas ligand in murine silicosis.[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fas ligand triggers pulmonary silicosis.
Borges VM
The Journal of Experimental Medicine, 194 (2), 155-164 (2001)
Eric Piaton et al.
Annales de pathologie, 35(4), 294-305 (2015-07-21)
May-Grünwald-Giemsa (MGG) stain is a Romanowsky-type, polychromatic stain as those of Giemsa, Leishman and Wright. Apart being the reference method of haematology, it has become a routine stain of diagnostic cytopathology for the study of air-dried preparations (lymph node imprints
Inflammatory response following acute magnesium deficiency in the rat.
Malpuech BC
Biochim. Biophys. Acta Gen. Subj., 1501 (2), 91-98 (2000)
Young-Whan Choi et al.
Journal of ethnopharmacology, 178, 17-24 (2015-12-18)
The giant butterbur Petasites japonicus is used to treat asthma and allergic diseases in traditional Korean, Japanese, and Chinese medicine. To elucidate the anti-allergic effect of Petasites genus, we studied effects of several compounds from Petasites japonicus leaves and found
Jin Huang et al.
Biomolecules & therapeutics, 26(6), 539-545 (2018-04-19)
4-Hydroxy-2-(4-hydroxyphenethyl)isoindoline-1,3-dione (PD1) is a synthetic phthalimide derivative of a marine compound. PD1 has peroxisome proliferator-activated receptor (PPAR) γ agonistic and anti-inflammatory effects. This study aimed to investigate the effect of PD1 on allergic asthma using rat basophilic leukemia (RBL)-2H3 mast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service