Skip to Content
Merck
All Photos(1)

Key Documents

00189

Sigma-Aldrich

Sulfadimidine solution

1 mg/mL in H2O

Synonym(s):

Sulfamethazine solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N4O2S
CAS Number:
Molecular Weight:
278.33
Beilstein:
261304
MDL number:
UNSPSC Code:
51283901
PubChem Substance ID:
Pricing and availability is not currently available.

form

liquid

Quality Level

concentration

1 mg/mL in H2O

color

colorless

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

enzyme | inhibits

SMILES string

Cc1cc(C)nc(NS(=O)(=O)c2ccc(N)cc2)n1

InChI

1S/C12H14N4O2S/c1-8-7-9(2)15-12(14-8)16-19(17,18)11-5-3-10(13)4-6-11/h3-7H,13H2,1-2H3,(H,14,15,16)

InChI key

ASWVTGNCAZCNNR-UHFFFAOYSA-N

Application

Sulfadimidine is used to induce cytochrome P450 3A4 (CYP3A4)[1] and to inhibit dihydrofolate reductase (DHFR). It was used to study amantadine acetylation[2].

Biochem/physiol Actions

Sulfadimidine is a competitive inhibitor of dihydropteroate synthetase to block the synthesis of folic acid. It acts as an inhibitor of dihydrofolate reductase (DHFR).

Packaging

10ML

Other Notes

Conditions for safe storage,including any incompatibilities Keep container tightly closed in a dry and well-ventilated place.Store under inert gas. Light sensitive.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A P Bras et al.
Drug metabolism and disposition: the biological fate of chemicals, 29(5), 676-680 (2001-04-17)
Amantadine acetylation was demonstrated to occur both in vivo and in vitro using transgenic male mice overexpressing spermidine/spermine N(1)-acetyltransferase (SSAT). We previously reported that neither NAT1 nor NAT2 was responsible for catalyzing acetylation of the primary amine group of amantadine.
Gang Luo et al.
Drug metabolism and disposition: the biological fate of chemicals, 30(7), 795-804 (2002-06-18)
Induction of cytochrome P450 3A4 (CYP3A4) is determined typically by employing primary culture of human hepatocytes and measuring CYP3A4 mRNA, protein and microsomal activity. Recently a pregnane X receptor (PXR) reporter gene assay was established to screen CYP3A4 inducers. To
Vincent Curtin et al.
Molecular pharmaceutics, 10(1), 386-396 (2012-11-29)
The coprocessing of active pharmaceutical ingredient (API) with an excipient which has a high glass transition temperature (T(g)) is a recognized strategy to stabilize the amorphous form of a drug. This work investigates whether coprocessing a model API, sulfadimidine (SDM)
Ma Jesús García-Galán et al.
The Science of the total environment, 409(24), 5505-5512 (2011-09-29)
Degradation of the sulfonamide sulfamethazine (SMZ) by the white-rot fungus Trametes versicolor was assessed. Elimination was achieved to nearly undetectable levels after 20 h in liquid medium when SMZ was added at 9 mg L(-1). Experiments with purified laccase and
M Rérat et al.
Preventive veterinary medicine, 103(4), 265-273 (2011-09-29)
The present study was conducted to evaluate the efficacy of two prophylactic antibiotic treatments against bovine respiratory disease (BRD) in veal calves. In addition, the antibiotic susceptibilities of isolated Pasteurellaceae were tested. The calves were treated either on the day

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service