Skip to Content
Merck
All Photos(1)

Documents

SMB00946

Sigma-Aldrich

Dihydrogenistein

≥95%

Synonym(s):

4′,5,7-Trihydroxyisoflavan-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O5
CAS Number:
Molecular Weight:
272.25
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25

biological source

synthetic

Quality Level

Assay

≥95%

form

solid

technique(s)

HPLC: suitable

color

white to yellow

storage temp.

2-8°C

InChI

1S/C15H12O5/c16-9-3-1-8(2-4-9)11-7-20-13-6-10(17)5-12(18)14(13)15(11)19/h1-6,11,16-18H,7H2

InChI key

UQGVUYNHDKMLSE-UHFFFAOYSA-N

Application

Dihydrogenistein is a metabolite of the soy isoflavone genistin.
Metabolomics research

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service