Skip to Content
Merck
All Photos(1)

Documents

RP-AH2-1

Recombinant Ara h 2 (Arachis hypogaea allergen 2), peanut

pkg of 250 μg, storage temp.:-20°C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.26

grade

analytical standard

Assay

>95%

mol wt

17-19 kDa (doublet)

packaging

pkg of 250 μg

manufacturer/tradename

InBio

technique(s)

HPLC: suitable
LC/MS: suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

General description

Produced and qualified by Indoor Biotechnologies.

Source: Pichia pastoris
Formulation: Preservative-free and carrier-free in phosphate buffered saline, 360mM NaCl, pH 7.4. Sterile filtered
Concentration: See product insert
Purification: Purified from Pichia pastoris culture by HPLC gel filtration. Purity > 95 % by silver stained SDS-PAGE

Note:
(1) rAra h 2 has a C-terminal 6xHis-tag.
(2) rAra h 2 appears as a doublet.
(3) Avoid repeated Freeze/Thaw cycles. It is recommended to aliquot the product upon first thaw and freeze in working aliquot volumes.

Disclaimer

Allergens are provided for research and commercial use in vitro:  not for human in vivo or therapeutic use.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification and characterization of a second major peanut allergen, Ara h II, with use of the sera of patients with atopic dermatitis and positive peanut challenge.
Burks AW.
The Journal of Allergy and Clinical Immunology, 90, 926-969 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service