NIST1624D
Sulfur in diesel fuel oil
NIST® SRM® 1624d, (0.4%)
Synonym(s):
Diesel fuel, Diesel fuel no. 2-D, Diesel oil, Diesel oil no. 2-D, Diesel oil; medium, Fuel oil #2
About This Item
Recommended Products
grade
certified reference material
Quality Level
manufacturer/tradename
NIST®
application(s)
petroleum
format
matrix material
Looking for similar products? Visit Product Comparison Guide
General description
SRM 1624D_cert
SRM 1624D_SDS
Application
Features and Benefits
- Certified mass fraction value is metrologically traceable to SI-derived unit.
- Details of expiration date, homogeneity, and storage, handling, and use instructions are provided on NIST certificate.
- Non-certified values of physical properties such as density, flash point, kinematic viscosity, oxidation stability, and carbon and hydrogen contents are also available on the certificate as well.
Other Notes
Sulfur (S)
Reference/Informational value is provided for the following analytes.
Carbon (C), Hydrogen (H)
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3
Target Organs
Central nervous system, thymus,Liver,Bone marrow
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
141.8 °F
Flash Point(C)
61 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service