Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of rod
manufacturer/tradename
NIST®
format
matrix material
Related Categories
General description
This Standard Reference Material (SRM) is intended for use in the evaluation of methods and the calibration of equipment used in the determination of oxygen in iron or ferrous materials. SRM 1090 is in the form of an ingot iron rod, 0.635 cm in diameter and 10.2 cm long (1/4 in. diamter and 4 in. long.) For more information, please refer to the COA and SDS.
SRM 1090_cert
SRM 1090_SDS
SRM 1090_cert
SRM 1090_SDS
Other Notes
Certified for the analytes listed below.
Oxygen (O)
Reference/Informational value is provided for the following analytes.
Nitrogen (N)
See certificate for values and more details at nist.gov/SRM.
Oxygen (O)
Reference/Informational value is provided for the following analytes.
Nitrogen (N)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service