Skip to Content
Merck
All Photos(1)

Documents

IRMM526B

Niobium

IRMM®, certified reference material, 0.1 mm foil

Synonym(s):

Columbium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Nb
CAS Number:
Molecular Weight:
92.91
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

IRMM®

manufacturer/tradename

JRC

resistivity

13-16 μΩ-cm, 20°C

bp

4742 °C (lit.)

mp

2468 °C (lit.)

density

8.57 g/mL at 25 °C (lit.)

application(s)

industrial hygiene
pharmaceutical (small molecule)

format

matrix material

SMILES string

[Nb]

InChI

1S/Nb

InChI key

GUCVJGMIXFAOAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
IRMM526B

Legal Information

IRMM is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christos D Malliakas et al.
Journal of the American Chemical Society, 135(5), 1719-1722 (2013-01-23)
2H-NbSe(2) is a canonical Charge-Density-Wave (CDW) layered material the structural details of which remained elusive. We report the detailed structure of 2H-NbSe(2) below the CDW transition using a (3 + 2)-dimensional crystallographic approach on single crystal X-ray diffraction data collected
Kulbir Kaur Ghuman et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(8), 085501-085501 (2013-01-25)
A systematic study of electronic structure and band gap states is conducted to analyze the monodoping and charge compensated codoping of rutile TiO(2) with Rh and Nb, using the DFT + U approach. Doping of rutile TiO(2) with Rh atoms induces hybridized
M I Tsindlekht et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(8), 085701-085701 (2013-01-31)
We report results of measurements of the ac conductivity of a Nb superconducting thin film in a swept dc magnetic field. In the mixed state the swept dc field creates vortices at the film surface which pass through the film
Tzyy-Jiann Wang et al.
Optics express, 20(27), 28119-28124 (2012-12-25)
We report ultra-smooth LiNbO(3) microdisk resonators fabricated by selective ion implantation, chemical etching, and thermal treatment. The undercut microdisk structure is produced by chemically etching the buried lattice damage layer formed by selective ion implantation. By thermal treatment, surface tension
Jason S Pelc et al.
Optics express, 20(25), 27510-27519 (2012-12-25)
Long-distance quantum communication networks require appropriate interfaces between matter qubit-based nodes and low-loss photonic quantum channels. We implement a downconversion quantum interface, where the single photons emitted from a semiconductor quantum dot at 910 nm are downconverted to 1560 nm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service