BP662
Methylphenobarbital
British Pharmacopoeia (BP) Reference Standard
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
methylphenobarbital
form
solid
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
BP
drug control
psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)
application(s)
pharmaceutical
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
SMILES string
O=C1NC(C(CC)(C2=CC=CC=C2)C(N1C)=O)=O
InChI
1S/C13H14N2O3/c1-3-13(9-7-5-4-6-8-9)10(16)14-12(18)15(2)11(13)17/h4-8H,3H2,1-2H3,(H,14,16,18)
InChI key
ALARQZQTBTVLJV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Other Notes
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service