Skip to Content
Merck
All Photos(1)

Documents

936715

Sigma-Aldrich

Grubbs Catalyst® M300 ChemBeads

new

Synonym(s):

Dichloro[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene](benzylidene)bis(3-bromopyridine)ruthenium(II) ChemBeads, Grubbs Catalyst® 3rd Generation ChemBeads, Grubbs Catalyst® C884 ChemBeads, Grubbs Catalyst® M37a (C884) ChemBeads, [1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro(phenylmethylene)bis(3-bromopyridine)ruthenium(II) ChemBeads, [1,3-Dimesityl-2-imidazolidinylidene]dichloro(phenylmethylene)bis(3-bromopyridine)ruthenium(II) ChemBeads

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H40Br2Cl2N4Ru
CAS Number:
Molecular Weight:
884.54
UNSPSC Code:
12352300

form

solid

Quality Level

composition

loading of catalyst, 4-6 wt. %

reaction suitability

reagent type: catalyst
core: ruthenium
reaction type: Ring-Opening Polymerization

storage temp.

2-8°C

SMILES string

Brc1cccnc1.Brc2cccnc2.Cc3cc(C)c(N4CCN(C4=[Ru](Cl)(Cl)=Cc5ccccc5)c6c(C)cc(C)cc6C)c(C)c3

InChI

1S/C21H26N2.C7H6.2C5H4BrN.2ClH.Ru/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;1-7-5-3-2-4-6-7;2*6-5-2-1-3-7-4-5;;;/h9-12H,7-8H2,1-6H3;1-6H;2*1-4H;2*1H;/q;;;;;;+2/p-2

InChI key

YDOACSUCKSODBO-UHFFFAOYSA-L

General description

ChemBeads glass beads coated with Grubbs Catalyst® M300.

Application

Metathesis: Ruthenium-Based Metathesis Catalysts Labile pyridine ligands result in a very fast initiator (even at 0 °C). Effective in cross metathesis of acrylonitrile. Also effective in RCM and the ROMP production of block copolymers and polymers with narrow PDIs

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

For larger scale uses, product also available in powdered form (682330)

Legal Information

Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service