93521
Bis[tris(3-(1H,1H,2H,2H-perfluorodecyl)phenyl)phosphine]palladium(II) dichloride
technical, ≥90% (AAS)
Synonym(s):
Bis[tris(3-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)phenyl)phosphine]palladium(II) dichloride
About This Item
grade
technical
Assay
≥90% (AAS)
form
powder
reaction suitability
reagent type: catalyst
core: palladium
mp
140-145 °C
SMILES string
Cl[Pd]Cl.FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCc1cccc(c1)P(c2cccc(CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)c2)c3cccc(CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)c3.FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCc4cccc(c4)P(c5cccc(CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)c5)c6cccc(CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)c6
InChI
1S/2C48H24F51P.2ClH.Pd/c2*49-25(50,28(55,56)31(61,62)34(67,68)37(73,74)40(79,80)43(85,86)46(91,92)93)13-10-19-4-1-7-22(16-19)100(23-8-2-5-20(17-23)11-14-26(51,52)29(57,58)32(63,64)35(69,70)38(75,76)41(81,82)44(87,88)47(94,95)96)24-9-3-6-21(18-24)12-15-27(53,54)30(59,60)33(65,66)36(71,72)39(77,78)42(83,84)45(89,90)48(97,98)99;;;/h2*1-9,16-18H,10-15H2;2*1H;/q;;;;+2/p-2
InChI key
HIDVEVKTXQUVSJ-UHFFFAOYSA-L
Application
Packaging
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1
Target Organs
Liver
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service