Skip to Content
Merck
All Photos(1)

Key Documents

88497

Supelco

3-Buten-2-ol

analytical standard

Synonym(s):

Methyl vinyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH(OH)CH3
CAS Number:
Molecular Weight:
72.11
Beilstein:
1719579
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.415 (lit.)

bp

96-97 °C (lit.)

density

0.832 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(O)C=C

InChI

1S/C4H8O/c1-3-4(2)5/h3-5H,1H2,2H3

InChI key

MKUWVMRNQOOSAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Buten-2-ol is a flavor and fragrance standard provided by Sigma-Aldrich®. Flavors and fragrances are widely used to add taste and/or smell to products without aroma, to mask unpleasant odors and to maintain stability of original flavor.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service