Skip to Content
Merck
All Photos(1)

Documents

87795

Sigma-Aldrich

2,3,4,5-Tetramethyl-2-cyclopentenone

technical, cis+trans, ≥90% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14O
CAS Number:
Molecular Weight:
138.21
Beilstein:
2324088
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥90% (GC)

refractive index

n20/D 1.476

density

0.927 g/mL at 20 °C (lit.)

SMILES string

CC1C(C)C(=O)C(C)=C1C

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service